Learn More
Cayman Chemical Ferritin PROTAC DeFer-2, Purity: ≥98%, 1 mg
SDP
Catalog No. 502916226
Encompass_Preferred
A PROTAC that drives ferritin degradation; induces ferritin degradation in B16/F10 cells at 0.5 µM; increases intracellular levels of free iron and induces pyroptosis in B16/F10 cells at 5 µM; ferritin PROTAC DeFer-2 encapsulated in albumin nanoparticles reduces tumor growth and increases survival in a B16/F10 murine melanoma model at 10 mg/kg
Catalog No. 50-291-6226 Supplier Cayman Chemical Supplier No. 389311 MG
A PROTAC that drives ferritin degradation; induces ferritin degradation in B16/F10 cells at 0.5 µM; increases intracellular levels of free iron and induces pyroptosis in B16/F10 cells at 5 µM; ferritin PROTAC DeFer-2 encapsulated in albumin nanoparticles reduces tumor growth and increases survival in a B16/F10 murine melanoma model at 10 mg/kg