Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ferrocene | 102-54-5 | 98.0% | C10H10Fe | 250 G
SDP

Catalog No. 5000358586
Encompass_Preferred

Ferrocene is a crucial structural core in bioorganometallic chemistry, recognized for its inherent stability, excellent redox properties, and low toxicity. This compound demonstrates antimalarial and anticancer properties, targeting parasites as part of the anti-infection pathway.

  • Inherent stability
  • Excellent redox properties
  • Low toxicity
  • Antimalarial properties
  • Anticancer properties
  • Targets parasites
  • Molecular formula: C10H10Fe
  • Molecular weight: 186.03

Catalog No. 50-003-58586 Supplier Medchemexpress LLC Supplier No. 250GHY79504
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ferrocene is a crucial structural core in bioorganometallic chemistry, recognized for its inherent stability, excellent redox properties, and low toxicity. This compound demonstrates antimalarial and anticancer properties, targeting parasites as part of the anti-infection pathway.

  • Inherent stability
  • Excellent redox properties
  • Low toxicity
  • Antimalarial properties
  • Anticancer properties
  • Targets parasites
  • Molecular formula: C10H10Fe
  • Molecular weight: 186.03

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.