Learn More
Medchemexpress LLC Ferrocene | 102-54-5 | MFCD00001427 | 98.0% | C10H10Fe | 100 G
SDP
Catalog No. 5000433123
Encompass_Preferred
Ferrocene is an important structural core in bioorganometallic chemistry due to its inherent stability, excellent redox properties, and low toxicity. It possesses antimalarial and anticancer properties.
  • Important structural core in bioorganometallic chemistry
  • Inherent stability
  • Excellent redox properties
  • Low toxicity
  • Possesses antimalarial properties
  • Possesses anticancer properties
Catalog No. 50-004-33123 Supplier Medchemexpress LLC Supplier No. HY79504100G
Ferrocene is an important structural core in bioorganometallic chemistry due to its inherent stability, excellent redox properties, and low toxicity. It possesses antimalarial and anticancer properties.
  • Important structural core in bioorganometallic chemistry
  • Inherent stability
  • Excellent redox properties
  • Low toxicity
  • Possesses antimalarial properties
  • Possesses anticancer properties