Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ferrocene

Catalog No. F4085G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
5 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F4085G 500 g
F408100G 5 g
F408500G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F4085G Supplier Sigma Aldrich Supplier No. F4085G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ferrocene

Specifications

CAS 102-54-5
Melting Point 172°C to 174°C
Boiling Point 249°C
Molecular Formula C10H10Fe
Linear Formula Fe(C5H5)2
MDL Number MFCD00001427
Quantity 500 g
Synonym Bis(cyclopentadienyl)iron; Di(cyclopentadienyl)iron
Molecular Weight (g/mol) 186.03 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.