Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrocene, 99%

Catalog No. p-7080356
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8720218 50 g
AA8720230 250 g
AA87202A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA8720218 Supplier Thermo Scientific Chemicals Supplier No. 08720218
Only null left
Add to Cart

CAS: 102-54-5 | C10H10Fe | 186.04 g/mol

Ferrocene is used as a catalyst for carbon nanotubes and for rocket propellants. It is also used in the fuel for petrol engines, gasoline antiknock, luminous sensitive materials and exhibit anticancer activity. It acts as a photosensitive catalyst and promote the photochemical degadation of plastic films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-54-5
Molecular Formula C10H10Fe
Molecular Weight (g/mol) 186.04
MDL Number MFCD00001427
InChI Key DFRHTHSZMBROSH-UHFFFAOYSA-N
Synonym ferrocene, bis cyclopentadienyl iron
PubChem CID 25199998
IUPAC Name cyclopenta-1,3-diene;iron
SMILES [Fe].c1cccc1.c1cccc1

Specifications

Melting Point 173°C to 174°C
Color Yellow to Orange
Boiling Point 249°C
Odor Characteristic
Quantity 50 g
UN Number UN1325
Sensitivity Air and moisture sensitive
Merck Index 14,4037
Solubility Information Insoluble in water. Soluble in organic solvents.
Formula Weight 186.04
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Ferrocene
Show More Show Less
Hazard Category H228-H302
Hazard Statement GHS H Statement
H228-H302
Flammable solid.
Harmful if swallowed.
Precautionary Statement P210-P240-P241-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber 203-039-3
RTECSNumber LK0700000
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.