Learn More
Strem, An Ascensus Company CAS# 12093-10-6. 10g. Ferrocene carboxaldehyde, min. 98%. MFCD00001429
SDP
Catalog No. 5090111085
Encompass
CAS# 12093-10-6. 10g. Ferrocene carboxaldehyde, min. 98%. MFCD00001429. Molecular Weight: 214.05. Molecular Formula: H(O)CC5H4FeC5H5. Color/form: orange to red xtl. Strem# 26-1750. http://www.strem.com/catalog/v/26-1750/
Catalog No. 50-901-11085 Supplier Strem, An Ascensus Company Supplier No. 26175010G
CAS# 12093-10-6. 10g. Ferrocene carboxaldehyde, min. 98%. MFCD00001429. Molecular Weight: 214.05. Molecular Formula: H(O)CC5H4FeC5H5. Color/form: orange to red xtl. Strem# 26-1750. http://www.strem.com/catalog/v/26-1750/