Learn More
Medchemexpress LLC Ferrocene-d10 | 12082-87-0 | 99.12% | C10D10Fe | 25 MG
SDP
Catalog No. 5000358010
Encompass_Preferred
Ferrocene-d10 is the deuterium labeled Ferrocene. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-58010 Supplier Medchemexpress LLC Supplier No. 25MGHY79504S
Ferrocene-d10 is the deuterium labeled Ferrocene. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.