Learn More
Medchemexpress LLC Ferrocene-d10 | 12082-87-0 | 99.12% | C10D10Fe | 50 MG
SDP
Catalog No. 5000433253
Encompass_Preferred
Ferrocene-d10 is a deuterium labeled Ferrocene intended for research use only. It can be utilized as a tracer and an internal standard for quantitative analysis, particularly in NMR, GC-MS, or LC-MS applications. Deuterium substitution may impact the pharmacokinetic and metabolic profiles of drugs. This product has a molecular weight of 196.09 and appears as a solid, typically light yellow to brown in color.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Features deuterium labeling
  • Solid form with a light yellow to brown appearance
Catalog No. 50-004-33253 Supplier Medchemexpress LLC Supplier No. HY79504S50MG
Ferrocene-d10 is a deuterium labeled Ferrocene intended for research use only. It can be utilized as a tracer and an internal standard for quantitative analysis, particularly in NMR, GC-MS, or LC-MS applications. Deuterium substitution may impact the pharmacokinetic and metabolic profiles of drugs. This product has a molecular weight of 196.09 and appears as a solid, typically light yellow to brown in color.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Features deuterium labeling
  • Solid form with a light yellow to brown appearance