Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrocenoyl chloride, For HPLC derivatization, ≥96.0% (AT), MilliporeSigma™ Supelco™

Catalog No. 111018057 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8057 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8057 Supplier MilliporeSigma Supelco Supplier No. 910321G
Add to Cart
Add to Cart

Chemical Identifiers

Synonym 1-Chlorocarbonylferrocene

Specifications

CAS 1293-79-4
Linear Formula C11H9 ClFeO
Quantity 1 g
Formula Weight 248.49
Percent Purity ≥96.0% (AT)
Grade For HPLC derivatization
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.