Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Ferrocenylmethyl)dodecyldimethylammonium Bromide 97.0+%, TCI America™
SDP

Catalog No. F02865G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F02865G Supplier TCI America Supplier No. F02865G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 98778-40-6
Molecular Formula C25H42BrFeN
MDL Number MFCD00144308
Synonym Dodecyl(ferrocenylmethyl)dimethylammonium Bromide

Specifications

Melting Point 137°C
Quantity 5 g
Formula Weight 492.37
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (Ferrocenylmethyl)dodecyldimethylammonium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.