Learn More
Medchemexpress LLC Ferrocenylmethyltrimethylammonium iodide | 12086-40-7 | MFCD00041534 | >98.5% | 385.08 g·mol^-1 | C14H20FeIN | 250 MG
SDP
Catalog No. 5000209492
Encompass_Preferred
Ferrocenylmethyltrimethylammonium iodide is an organometallic quaternary ammonium salt used as a biochemical reagent and synthetic intermediate in organic and materials research. It is a crystalline iodide salt with molecular formula C14H20FeIN and molecular weight about 385.08 g·mol^-1; common applications include use as a reagent, catalyst, and precursor for organometallic synthesis and electrochemical studies.
  • High purity reagent suitable for research applications.
  • Useful as a reagent and phase-transfer catalyst in organic synthesis.
  • Serves as a synthetic intermediate for organometallic compounds.
  • Useful for electrochemical and materials research because of ferrocenyl redox properties.
  • Store protected from light at 4°C; in solution, store at -80°C for long-term stability.
  • Typically supplied in 250 mg laboratory quantities.
Catalog No. 50-002-09492 Supplier Medchemexpress LLC Supplier No. 250MGHYW075836
Ferrocenylmethyltrimethylammonium iodide is an organometallic quaternary ammonium salt used as a biochemical reagent and synthetic intermediate in organic and materials research. It is a crystalline iodide salt with molecular formula C14H20FeIN and molecular weight about 385.08 g·mol^-1; common applications include use as a reagent, catalyst, and precursor for organometallic synthesis and electrochemical studies.
  • High purity reagent suitable for research applications.
  • Useful as a reagent and phase-transfer catalyst in organic synthesis.
  • Serves as a synthetic intermediate for organometallic compounds.
  • Useful for electrochemical and materials research because of ferrocenyl redox properties.
  • Store protected from light at 4°C; in solution, store at -80°C for long-term stability.
  • Typically supplied in 250 mg laboratory quantities.