Learn More
Qc Supply LLC FERRODEX IRON DEXTRAN 100ML
SDP
Catalog No. NC2177014
Encompass_Preferred
agrilabsIron Dextran treats and prevents anemia due to iron deficiency in baby pigs.Directions/Dosage: 1 mL IM 2 - 4 days of age.Contents: 100 mL bottle
Catalog No. NC2177014 Supplier Qc Supply LLC Supplier No. 540565
agrilabsIron Dextran treats and prevents anemia due to iron deficiency in baby pigs.Directions/Dosage: 1 mL IM 2 - 4 days of age.Contents: 100 mL bottle