Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Ferrofluid
GREENER_CHOICESCI_ED

Catalog No. S37729 Shop All Aldon Products
Change view
Click to view available options
Class Size:
1 Class
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S37729 1 Class
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This is not a toy, this product is intended for scientific use by ages 13 and up. DOT Info: Non regulated
Catalog No. S37729 Supplier Innovating Science™ Supplier No. IS2801
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This is not a toy, this product is intended for scientific use by ages 13 and up. DOT Info: Non regulated

Includes

50 mL Ferrofluid

Demonstrate magnetic fields to your students in a very visual way.

Ferrofluid is a colloidal liquid made of particles less than 10 nanometers in diameter. When exposed to a magnetic field, the nanoparticles form regular patterns of peaks and valleys.

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Demonstrate magnetic fields to students in a very visual way.
Includes 50 mL Ferrofluid
Quantity 50 mL Ferrofluid
Dimensions (L x W x H) 7 x 6 x 5 in.
Class Size 1 Class
Product Type Ferrofluid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.