Learn More
FERROTEC USA CORPORATION FERROFLUIDICS ADVENTRE SCI KIT
Supplier:  FERROTEC USA CORPORATION 92EK1000000
Encompass_Preferred
Perform a variety of experiments and demonstrations using liquid-manipulating magnets. Kit, Educational; Ferrofluid Adventure Science Kit; Assembled teaching tool; To visualize magnetic patterns; For students of all grade levels. Includes: 50 cc ferrofluid; transfer pipets; syringe; display cell; blue button magnets; ring magnet; aluminum dishes; latex gloves; instruction booklet, 17 pages.
Catalog No. NC1083323