Learn More
Hach Company Ferroin Indicator Solution, 25 mL DB
Supplier:  Hach Company 2055133
Encompass
Indicator for Low Range COD determination by ferrous titration. APHA formulation
Catalog No. 50-199-5189