Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferroin Indicator, 0.025M, Solution, Spectrum™ Chemical
SDP

Catalog No. 18610550 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Catalog No. Quantity
18-610-550 500 mL
18-610-549 100 mL
2 options

Catalog No. 18-610-550

Supplier: Spectrum Chemical Manufacturing Corporation F157500MLGL

Only null left
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Ferroin Indicator Solution
CAS 7782-63-0, 5144-89-8, 7732-18-5
Concentration 0.025 M
Grade Chemical Solution
Packaging Amber Glass Bottle
Color Dark Red
Quantity 500 mL

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.