Learn More
Aqua Solutions FERROIN INDICATOR .025 M 100ML
SDP
Catalog No. NC9122730
Encompass_Preferred
Ferroin Indicator; 0.025 M; Redox Indicator; Dark red odorless liquid; 100mL; M.P. ~0 deg.C; B.P. ~100 deg.C; Specific Gravity: ~1; Wash thoroughly after handling
Catalog No. NC9122730 Supplier Aqua Solutions Supplier No. 3820100ML
Ferroin Indicator; 0.025 M; Redox Indicator; Dark red odorless liquid; 100mL; M.P. ~0 deg.C; B.P. ~100 deg.C; Specific Gravity: ~1; Wash thoroughly after handling