Learn More
Hach Company Ferroin Indicator Solution, 29 mL DB
Supplier:  Hach Company 181233
Encompass
Oxidation-reduction indicator solution used in determination of COD by ferrous titration.
Catalog No. 50-199-4606