Learn More
Med Vet International Ferromatic Automatic Syringe, J0053FM18, Each
Catalog No. 5000582328
Encompass_Preferred
Unique design has dosage adjustments from .5ml to 5.0ml. Fits bottle tops from 30ml–120ml. • One-piece front end with luer lock J0053fm18 NOTE: The following parts are shown on the parts diagram, but are not sold individually. They must be purchased as part of a kit: J0053fmd31 (10 x 2, 11 x 2, 12 x 2, 14, 20, 21, 24, 25, 26).
Catalog No. 50-005-82328 Supplier Med Vet International Supplier No. JOR53FM18
Unique design has dosage adjustments from .5ml to 5.0ml. Fits bottle tops from 30ml–120ml. • One-piece front end with luer lock J0053fm18 NOTE: The following parts are shown on the parts diagram, but are not sold individually. They must be purchased as part of a kit: J0053fmd31 (10 x 2, 11 x 2, 12 x 2, 14, 20, 21, 24, 25, 26).