Learn More
Hach Company FerroMo™ Iron Reagent 1 Powder Pillows, pk/25
Supplier:  Hach Company 2543768
Encompass
For determination of iron in cooling waters containing molybdate-based treatments. Hach Method 8365. Range: 0.01 - 1.80 mg/L Fe. Pack of 25 powder pillows
Catalog No. 50-199-6199