Learn More
Hach Company FerroMo™ Iron Reagent 2 Powder Pillows, pk/50
Supplier: Hach Company 2543866
For determination of iron in cooling waters containing molybdate-based treatments. Hach Method 8365. Range: 0.01 - 1.80 mg/L Fe. Pack of 50 powder pillows