Learn More
Biomatik Corporation Ferroportin (Fpn) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29504
Encompass_Preferred
Ferroportin (Fpn) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Ferroportin (FPN). The Ferroportin (Fpn) Monoclonal Antibody has been validated for the following applications: WB. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Trp127~Val321. Applications: WB. Target Synonyms: Ferroportin (FPN). Accession Number: Q9NP59. Gene ID: 30061. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4831