Learn More
Cayman Chemical FerroprotoporphyrIn 100mg
SDP
Catalog No. 502946784
Encompass_Preferred
An iron-containing cofactor; involved the transport or storage of oxygen by hemoglobin or myoglobin, respectively, electron transfer by cytochrome b5, and oxidation of xenobiotics or endogenous substrates by CYPs
Catalog No. 50-294-6784 Supplier Cayman Chemical Supplier No. 34327100 MG
An iron-containing cofactor; involved the transport or storage of oxygen by hemoglobin or myoglobin, respectively, electron transfer by cytochrome b5, and oxidation of xenobiotics or endogenous substrates by CYPs