Learn More
Cayman Chemical FerroptosIs Inducer 24 10mg
SDP
Catalog No. 502944811
Encompass_Preferred
An inducer of ferroptosis; inhibits GPX4 by 30-35% at 50 µM; increases the levels of HO-1 in MDA-MB-231 cancer cells, as well as increases HO-1 bilirubin formation activity in MDA-MB-231 cell lysates at 10 µM; reduces the viability of, as well as increases the levels of oxidized lipids, decreases the levels of IL-6 and GST, and disrupts the mitochondrial membrane potential in, MDA-MB-231 cells at 50 µM; increases the levels of ROS in a time-dependent manner
Catalog No. 50-294-4811 Supplier Cayman Chemical Supplier No. 4120110 MG
An inducer of ferroptosis; inhibits GPX4 by 30-35% at 50 µM; increases the levels of HO-1 in MDA-MB-231 cancer cells, as well as increases HO-1 bilirubin formation activity in MDA-MB-231 cell lysates at 10 µM; reduces the viability of, as well as increases the levels of oxidized lipids, decreases the levels of IL-6 and GST, and disrupts the mitochondrial membrane potential in, MDA-MB-231 cells at 50 µM; increases the levels of ROS in a time-dependent manner