Learn More
Medchemexpress LLC HY-19364 5mg Medchemexpress, Ferroquine CAS:185055-67-8 Purity:>98%
SDP
Catalog No. 502082126
Encompass_Preferred
Medchemexpress, HY-19364 5mg Ferroquine CAS:185055-67-8 Ferroquine (Ferrochloroquine), a ferrocenyl analogue of Chloroquine, is an antimalarial agent. Ferroquine shows parasiticidal effect on Plasmodium by inducing oxidative stress and the subsequent destruction of the membrane. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2126 Supplier Medchemexpress LLC Supplier No. HY193645MG
Medchemexpress, HY-19364 5mg Ferroquine CAS:185055-67-8 Ferroquine (Ferrochloroquine), a ferrocenyl analogue of Chloroquine, is an antimalarial agent. Ferroquine shows parasiticidal effect on Plasmodium by inducing oxidative stress and the subsequent destruction of the membrane. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.