Learn More
Medchemexpress LLC HY-100579 10mg , Ferrostatin-1 CAS:347174-05-4 Purity:>98%
SDP
Catalog No. 502259214
Encompass_Preferred
HY-100579 10mg Ferrostatin-1 CAS: 347174-05-4 Ferrostatin-1 (Fer-1), a potent and selective ferroptosis inhibitor, suppresses Erastin-induced ferroptosis in HT-1080 cells (EC50=60 nM). Ferrostatin-1, a synthetic antioxidant, acts via a reductive mechanism to prevent damage to membrane lipids and thereby inhibits cell death. Antifungal Activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9214 Supplier Medchemexpress LLC Supplier No. HY10057910MG
HY-100579 10mg Ferrostatin-1 CAS: 347174-05-4 Ferrostatin-1 (Fer-1), a potent and selective ferroptosis inhibitor, suppresses Erastin-induced ferroptosis in HT-1080 cells (EC50=60 nM). Ferrostatin-1, a synthetic antioxidant, acts via a reductive mechanism to prevent damage to membrane lipids and thereby inhibits cell death. Antifungal Activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.