Learn More
Cayman Chemical FerrostatIn-1 DIyn 10mg
SDP
Catalog No. 503015897
Encompass_Preferred
A ferroptosis inhibitor; inhibits erastin-induced cell death in HT-1080 cells from 0.01-1 μM; contains a diyne small vibrational tag and has been used to assess localization of ferrostatin-1 in HT-1080 cells using stimulated Ramen scattering microscopy
Catalog No. 50-301-5897 Supplier Cayman Chemical Supplier No. 2549210 MG
A ferroptosis inhibitor; inhibits erastin-induced cell death in HT-1080 cells from 0.01-1 μM; contains a diyne small vibrational tag and has been used to assess localization of ferrostatin-1 in HT-1080 cells using stimulated Ramen scattering microscopy