Learn More
Apexbio Technology LLC Ferrostatin-1 (Fer-1), 347174-05-4, MFCD08072959, 5 mg
SDP
Supplier:  Apexbio Technology LLC A43715
Encompass_Preferred
MF: C15H22N2O2, MW: 262.35, Solubility: 9.8 mg/mL in DMSO, 99.6 mg/mL in EtOH with ultrasonic,insoluble in H2O. Ferrostatin-1 is a selective inhibitor of ferroptosis.
Catalog No. 50-101-2713