Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Ammonium Sulfate, 0.1N Mohr's Salt Solution, Reagents

Catalog No. p-200021654
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-711 1 L
01-334-709 4 L
01-334-708 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-334-711 Supplier Reagents Holdings Llc Supplier No. CM1297001A
Only null left
Add to Cart
Add to Cart

Ferrous Ammonium Sulfate, 0.1N Mohr's Salt Solution, Requires Periodic Restandardization, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Specifications

Chemical Name or Material Ferrous Ammonium Sulfate, 0.1N Mohr's Salt Solution, Requires Periodic Restandardization
Boiling Point 100°C
Color Light Green
CAS 7732-18-5, 7664-93-9
pH <1
Physical Form Liquid
Quantity 1 L
Synonym Iron(II) Ammonium Sulfate
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.05
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.