Learn More
Hach Company Ferrous Ammonium Sulfate, Hexahydrate, 113 g
Catalog No. 501994845 Shop All Hach Company Products
Encompass
ACS Grade, CAS No. 7783-85-9 Fe(NH4)2(SO4)2·...6H2O, FW = 392.14. Assay: 98.5 - 101.5 g
Catalog No. 50-199-4845 Supplier Hach Company Supplier No. 1125614
ACS Grade, CAS No. 7783-85-9 Fe(NH4)2(SO4)2·...6H2O, FW = 392.14. Assay: 98.5 - 101.5 g