Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Ammonium Sulfate ACS AR Crystal, Macron Fine Chemicals™

Catalog No. MK5064212 Shop All Avantor Products
Change view
Click to view available options
Poly; 2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-5064-212 Poly; 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-5064-212 Supplier Avantor Supplier No. 506406

Specifications

CAS 7783-85-9
Physical Form Crystalline
Quantity 2.5 kg
Molecular Formula FeH20N2O14S2
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
IUPAC Name λ²-iron(2+) diammonium hexahydrate disulfate
Molecular Weight (g/mol) 392.13
Formula Weight 392.13
Grade ACS
Packaging Poly
Chemical Name or Material Ferrous Ammonium Sulfate, 6-Hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.