Learn More
Hach Company Ferrous Ammonium Sulfate Sol.
Supplier:  Hach Company 181133NC
Encompass
Ferrous Ammonium Sulfate Solution 0 25 N-29 mLFor sample dechlorination 29 mL dropping bottle
Catalog No. 50-002-06981