Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Ammonium Sulfate Solution (1%), ENG Scientific

Catalog No. ES9870
Change view
Click to view available options
Quantity:
1 x 3800 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES9870 1 x 3800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ES9870 Supplier Eng Scientific Inc Supplier No. 9870
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ferrous Ammonium Sulfate Solution (1%), ENG Scientific

Specifications

Packaging Bottle
Quantity 1 x 3800 mL
Chemical Name or Material Ferrous Ammonium Sulfate Solution, 1%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.