Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Ammonium Sulfate Titrant, For Chlorine, Chlorine Dioxide (1mL = 0.1mg Cl), Certified, 0.00282N ±0.00010N (0.00282M), LabChem™

Catalog No. LC145501
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC145501 500 mL
LC145502 1 L
LC145504 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC145501 Supplier LabChem, Inc. Supplier No. LC145501
Add to Cart
Add to Cart

Specifications

Color Clear pale green
Concentration 0.00282N ±0.00010N (0.00282M)
Format Liquid
Packaging Amber Glass
Quantity 500 mL
Storage Requirements Room Temperature
Type Ferrous Ammonium Sulfate Titrant
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.