Learn More
Medchemexpress LLC Ferrous bisglycinate | 20150-34-9 | 98.0% | 100 G
SDP
Catalog No. 5000340495
Encompass_Preferred
Ferrous bisglycinate is an orally active iron fortificant and therapeutic iron supplement. It can be used for the research of iron deficiency anemia.
  • Does not affect Caco-2 cells viability (at 25-200 μM).
  • Increases ferritin content in Caco-2 cells.
  • Exerts a protective effect on colitis in mice.
  • Demonstrated best survival rates (100%) in a colitis animal model.
  • Caused the least body loss (9%) in a colitis animal model.
Catalog No. 50-003-40495 Supplier Medchemexpress LLC Supplier No. HY130078100G
Ferrous bisglycinate is an orally active iron fortificant and therapeutic iron supplement. It can be used for the research of iron deficiency anemia.
  • Does not affect Caco-2 cells viability (at 25-200 μM).
  • Increases ferritin content in Caco-2 cells.
  • Exerts a protective effect on colitis in mice.
  • Demonstrated best survival rates (100%) in a colitis animal model.
  • Caused the least body loss (9%) in a colitis animal model.