Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ferrous bisglycinate | 20150-34-9 | 98% | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHY130078

Encompass_Preferred

Ferrous bisglycinate is an orally active iron fortificant and therapeutic iron supplement that can be used for the research of iron deficiency anemia.

  • Orally active iron fortificant.
  • Therapeutic iron supplement.
  • Used for the research of iron deficiency anemia.
  • Molecular formula: C4H8FeN2O4.
  • Molecular weight: 203.96.
  • Target: Transferrin Receptor.
  • Pathway: Apoptosis.
  • Does not affect Caco-2 cells viability at specific concentrations and incubation times.
  • Increases ferritin content in Caco-2 cells.
  • Protective effect on colitis in mice.

Catalog No. 50-003-42082


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.