Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Chloride Tetrahydrate (Crystalline/Certified), Fisher Chemical

Catalog No. I90500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
I90-500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. I90-500

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13478-10-9 | Cl2FeH8O4 | 198.805 g/mol

Glass Bottle; 500g

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ferrous Chloride Tetrahydrate
Melting Point 105°C
Color Green
CAS 13478-10-9
pH 2.5
Physical Form Solid
Quantity 500 g
Assay Percent Range 99 to 102%
Molecular Formula Cl2FeH8O4
MDL Number MFCD00149709
UN Number NA 1759
Synonym ferrous chloride tetrahydrate, iron ii chloride tetrahydrate, iron dichloride tetrahydrate, cl2fe.4h2o, fecl2.4h2o, iron chloride, tetrahydrate, dichloroiron-water 1/4, iron ii chloride tetra hydrate, ae>> nciu eae r masculineiii
Solubility Information Other solubilities: Soluble in alcohol
InChI Key WSSMOXHYUFMBLS-UHFFFAOYSA-L
SMILES O.O.O.O.Cl[Fe]Cl
IUPAC Name dichloroiron;tetrahydrate
Molecular Weight (g/mol) 198.805
PubChem CID 16211588
ChEBI CHEBI:86249
Formula Weight 198.81
Percent Purity 99 to 102%
Grade Certified
Identification Pass test
Packaging Glass Bottle
Density 1.93g/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.