Learn More
Hach Company Ferrous Ethylenediammonium Sulfate Digital Titrator Cartridge, 0.00564 N
Catalog No. 501995744 Shop All Hach Company Products
Encompass
For chlorine determination by Digital Titrator titration, using DPD Free or Total Chlorine Reagent indicator. Hach Method 8210. Range: Up to 3.00 mg/L Cl2. 0.00564 N FEAS. Digits x 0.01 = mg/L Cl2 in a 25 mL sample.
Catalog No. 50-199-5744 Supplier Hach Company Supplier No. 2292301
For chlorine determination by Digital Titrator titration, using DPD Free or Total Chlorine Reagent indicator. Hach Method 8210. Range: Up to 3.00 mg/L Cl2. 0.00564 N FEAS. Digits x 0.01 = mg/L Cl2 in a 25 mL sample.