Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Fluoborate, 40% Soln, MP Biomedicals

Catalog No. ICN222361 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN222361 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN222361 Supplier MP Biomedicals, Inc Supplier No. 0522236180

Chemical Identifiers

CAS 15283-51-9
Molecular Formula B2F8Fe
Molecular Weight (g/mol) 229.452
InChI Key GXZXLFFLHJJJLX-UHFFFAOYSA-N
Synonym iron ii tetrafluoroborate, ferrous fluoborate, iron bis tetrafluoroborate, fe bf4 2, iron 2+ ditetrafluoroborate, ksc005q1l, iron ion 2+ ditetrafluoroborate, ferrous fluoborate soln
PubChem CID 11736352
IUPAC Name iron(2+);ditetrafluoroborate
SMILES [B-](F)(F)(F)F.[B-](F)(F)(F)F.[Fe+2]

Specifications

Quantity 100 g
Chemical Name or Material Ferrous Fluoborate, 40% Soln
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.