Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Fumarate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013064 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3064 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3064 Supplier MilliporeSigma Supelco Supplier No. PHR14821G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ferrous fumarate is a commonly used inexpensive substitute for other forms of iron, that is employed as a food iron fortificant.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 141-01-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C4H2 FeO4
MDL Number MFCD00058315
Synonym Iron(II) fumarate; Ferrous fumarate
Type Certified Reference Material
Quantity 1 g
Formula Weight 169.90
Grade Certified Reference Material
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.