Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ferrous gluconate | 299-29-6 | MFCD00075298 | 99.3% | 446.14 | C12H22FeO14 | 100 G
SDP

Catalog No. 5000284169
Encompass_Preferred

Ferrous gluconate is an orally active iron supplement provided as a research-grade reagent for studies of iron-deficiency diseases. It is supplied as a solid reagent suitable for analytical and experimental applications, with documented solubility and storage recommendations to support laboratory use.

  • Orally active iron supplement for research of iron-deficiency diseases.
  • Research-grade solid reagent for analytical and experimental use.
  • Molecular formula C12H22FeO14; molecular weight 446.14.
  • Soluble in DMSO at 100 mg/mL; may require ultrasonic agitation.
  • Typical purity 99.25%.
  • Recommended storage: 4°C under nitrogen; in solution -80°C up to 6 months, -20°C up to 1 month.

Catalog No. 50-002-84169 Supplier Medchemexpress LLC Supplier No. HYW698249100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ferrous gluconate is an orally active iron supplement provided as a research-grade reagent for studies of iron-deficiency diseases. It is supplied as a solid reagent suitable for analytical and experimental applications, with documented solubility and storage recommendations to support laboratory use.

  • Orally active iron supplement for research of iron-deficiency diseases.
  • Research-grade solid reagent for analytical and experimental use.
  • Molecular formula C12H22FeO14; molecular weight 446.14.
  • Soluble in DMSO at 100 mg/mL; may require ultrasonic agitation.
  • Typical purity 99.25%.
  • Recommended storage: 4°C under nitrogen; in solution -80°C up to 6 months, -20°C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.