Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ferrous gluconate | 299-29-6 | MFCD00075298 | 99.3% | 446.14 | C12H22FeO14 | 25 G
SDP

Catalog No. 5000283946
Encompass_Preferred

Ferrous gluconate is an orally active iron supplement used as a research reagent in studies of iron-deficiency disorders and for analytical applications.

  • Orally active iron supplement for research applications.
  • High purity (99.3%).
  • Soluble in DMSO at 100 mg/mL with ultrasonic assistance.
  • Available in multiple laboratory pack sizes.
  • Storage: 4°C under nitrogen; in solution, -80°C for 6 months or -20°C for 1 month (under nitrogen).
  • Intended for research use only; not for human consumption.

Catalog No. 50-002-83946 Supplier Medchemexpress LLC Supplier No. HYW69824925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ferrous gluconate is an orally active iron supplement used as a research reagent in studies of iron-deficiency disorders and for analytical applications.

  • Orally active iron supplement for research applications.
  • High purity (99.3%).
  • Soluble in DMSO at 100 mg/mL with ultrasonic assistance.
  • Available in multiple laboratory pack sizes.
  • Storage: 4°C under nitrogen; in solution, -80°C for 6 months or -20°C for 1 month (under nitrogen).
  • Intended for research use only; not for human consumption.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.