Learn More
Hach Company Ferrous Iron Reagent Powder Pillows, pk/100
Supplier:  Hach Company 103769
Encompass
For determination of ferrous iron by the 1,10-phenanthroline method. Hach method 8146. Range: 0.02-3.00 mg/L Fe. Pack of 100 powder pillows.
Catalog No. 50-199-4494