Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Sulfate Heptahydrate, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCF037025F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCF037025F 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCF037025F Supplier Ricca Chemical Company Supplier No. RDCF037025F4
Add to Cart
Add to Cart
Request a Quote

(Iron (II) Sulfate Heptahydrate)

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
IUPAC Name λ2-iron(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 100
Physical Form Fine Crystals or Crystalline Powder
Quantity 25 kg
Percent Purity min 99%
Grade ACS
Packaging Fiber Drum
Chemical Name or Material Ferrous Sulfate Heptahydrate
ShelfLife 84 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.