Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Sulfate Heptahydrate, ACS Reagent (Iron (II) Sulfate Heptahydrate), Reagents

Catalog No. 01337957
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-957 500 g
01-337-961 100 g
01-337-959 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-957 Supplier Reagents Holdings Llc Supplier No. C1207000500A5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ferrous Sulfate Heptahydrate, ACS Reagent (Iron (II) Sulfate Heptahydrate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
Synonym Iron(II) Sulfate Heptahydrate
IUPAC Name λ²-iron(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

Boiling Point >300°C
Color Green or Bluish-Green
Physical Form Solid
Quantity 500 g
Percent Purity 99%
Grade ACS
Packaging Poly Bottle
Specific Gravity 1.897
Chemical Name or Material Ferrous Sulfate Heptahydrate, ACS Reagent (Iron (II) Sulfate Heptahydrate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.