Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ferrous Sulfide
SCI_ED

Catalog No. S25327 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Solid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25327 500 g Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25327
Add to Cart
Add to Cart

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 1317-37-9
Molecular Formula FeS
Molecular Weight (g/mol) 87.91
MDL Number MFCD00011013
InChI Key GNVXPFBEZCSHQZ-UHFFFAOYSA-N
Synonym ferrous sulfide, iron ii sulfide, iron sulfide, troilite, iron sulfuret, iron sulphide, iron monosulfide, iron protosulfide, ferrous monosulfide
PubChem CID 14828
IUPAC Name λ²-iron(2+) sulfanediide
SMILES [S--].[Fe++]

Specifications

Quantity 500 g
Linear Formula FeS
Formula Weight 87.91
Packaging Solid
Density 4.84g/mL
Chemical Name or Material Ferrous Sulfide
Safety handling Check Alert storage code Red
ShelfLife Two years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.