Learn More
Hach Company FerroVer Iron Reagent Accuvac Ampules, pk/25
Supplier:  Hach Company 2507025
Encompass
For determination of total dissolved Iron, as well as complexed iron compounds, by the 1,10 Phenanthroline Method - using FerroVer total iron reagent. Hach Method 8008. USEPA approved for reporting wastewater analysis results after acid digestion. Range: 0.02 - 3.00 mg/L Fe (0-5 mg/L with Pocket Colorimeter II). Pack of 25 ampules.
Catalog No. 50-199-6109