Learn More
Hach Company FerroVer Iron Reagent Powder Pillows, 10 mL, pk/100
Supplier:  Hach Company 2105769
Encompass
For determination of Iron by the 1,10 phenanthroline method. USEPA approved method for reporting wastewater analysis using Hach Method 8008. The FerroVer method recovers total soluble ferrous and ferric iron, as well as complexed iron compounds and many insoluble iron forms. Range: 0.02 to 3.00 mg/L. Sample Size: 10 mL Pack of 100 powder pillows. To confirm accurate results, see appropriate standard solutions on the Consumables tab.
Catalog No. 50-199-5399