Learn More
Hach Company FerroVer Iron Reagent Powder Pillows, 25 mL, pk/100
Supplier:  Hach Company 85499
Encompass
For Iron determination by the 1,10 phenanthroline method, using FerroVer Iron Reagent. Hach Method 8008. Sample size: 25 mL. USEPA approved for reporting wastewater analysis. Pack of 100 Powder Pillows To confirm accurate results, see appropriate standard solutions on the Consumables tab.
Catalog No. 50-199-4459