Learn More
Hach Company FerroVer Iron Reagent Powder Pillows, 5 mL, pk/50
Supplier: Hach Company 92746
For Iron determination by the 1,10 phenanthroline method, using FerroVer Iron Reagent Powder Pillows. For 5-mL samples. Pack of 50 Powder Pillows